Order Entry
Italy
ContactUsLinkComponent
Ethanol absolute for histology, J.T.Baker®
Flammable
Ethanol absolute for histology, J.T.Baker®
Supplier:  Avantor
Certificates
Flammable
Ethanol absolute for histology, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 99.8 %
  • Calcium (Ca):
    max. 5 ppm
  • Iron (Fe):
    max. 1 ppm
  • Residue after Evaporation:
    max. 0.0015 %
  • Substances Darkened by H2SO4:
    passes test
  • Titrable Acid (meq/g):
    max. 0.001
  • Water (H2O):
    max. 0.2 %

Specification Test Results

Related Information

Frequently Bought Together