Order Entry
Italy
Orders LinkContactUsLinkComponent
Ethanol, BAKER, Laboratory Reagent, J.T.Baker®

Ethanol, BAKER, Laboratory Reagent, J.T.Baker®

Flammable
Supplier:   Avantor
Ethanol, BAKER, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Ethanol, BAKER, Laboratory Reagent, J.T.Baker®

2 Options Available Below

Product Details & Documents
Synonyms:  Ethyl alcohol
  • CAS Number:64-17-5
  • Formula:H₃CCH₂OH
  • MDL Number:MFCD00003568
  • Molecular Weight:46.07 g/mol
  • Melting Pt:–114 °C
  • Storage Temperature:Ambient
  • EINECS:200-578-6
  • UN:1170
  • ADR:3,II

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Assay:
    96.0 - 97.2 % (v/v)
  • Appearance:
    passes test
  • Boiling Point:
    78.0 - 79.0 °C
  • Density (g/ml) at 20°C:
    0.804 - 0.809
  • Furfural:
    passes test
  • Fuseloil, aldehyde:
    passes test
  • Heavy Metals, Zinc (as Pb):
    max. 2 ppm
  • Identification (by IR):
    passes test
  • Iron (Fe):
    max. 1 ppm
  • Methanol (CH3OH):
    max. 500 ppm
  • Residue after Evaporation:
    max. 0,0015 %
  • Substances Reducing KMnO4:
    passes test
  • Titrable acid:
    passes test
  • Titrable base:
    passes test
  • UV-absorbance (1cm vs hexane):
    -
  • at 220 nm:
    max. 0,30
  • at 230 nm:
    max. 0,18
  • at 240 nm :
    max. 0,08
  • at 270 nm:
    max. 0,02
  • 220-270 nm (no maxima):
    passes test
Product Family Options

Product Information

  • Pack type
    Duty status
    Pk
    Availability
    Price
  • Plastic bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 100,33 €
    100,33 €
    1 * 2,5 L
       100,33 €

  • HDPE bottle
    Duty paid
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 100,33 €
    100,33 €
    1 * 2,5 L
       100,33 €