1-Hexanol 98%, Reagent Grade
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:111-27-3
:
Formula:H₃C(CH₂)₄CH₂OH
MDL Number:MFCD00002982
:102.18 g/mol
Merck Index:13,04715
Boiling Pt:11.8 %(V)
Melting Pt:–52 °C
Density:0.8 g/cm³ (20 °C)
Flash Pt:59 °C
Storage Temperature:Ambient
EINECS:203-852-3
UN:2282
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 97.5 %
- APHA:Max. 20
- Vapor Density:4.5 (vs air)
Specification Test Results
Frequently Bought Together