
1-Hexanol
Catalog # EHERC14196700
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:102.18 g/mol
Merck Index:13,04715
Boiling Pt:156 °C (1013 hPa)
Melting Pt:–45 °C
Density:0.8322 g/cm³ (20 °C)
Flash Pt:60 °C
Storage Temperature:Ambient
EINECS:203-852-3
UN:2282
ADR:3,III
:GHS07
CAS Number:111-27-3
Formula:H₃C(CH₂)₄CH₂OH
MDL Number:MFCD00002982