Order Entry
Italy
ContactUsLinkComponent
1-Decanol analytical standard, Supelco®
Irritant
Environment
1-Decanol analytical standard, Supelco®
Catalog # 442539.
Supplier:  Merck
CAS Number:  
undefined
Irritant
Environment
1-Decanol analytical standard, Supelco®
Catalog # 442539.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :158.28 g/mol
Merck Index:13,02875
Boiling Pt:230 °C (1013 hPa)
Melting Pt:7 °C
Density:0.82634 g/cm³ (20 °C)
Flash Pt:82 °C
Storage Temperature:Ambient
EINECS:203-956-9
UN:3082
ADR:9,III
 :GHS07
 :GHS09
CAS Number:112-30-1
Formula:H₃C(CH₂)₈CH₂OH
MDL Number:MFCD00004747