


Acetic anhydride ≥96%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:108-24-7
:
Formula:(CH₃CO)₂O
MDL Number:MFCD00008705
Merck Index:13,00057
Boiling Pt:139.9 °C (1013 hPa)
Melting Pt:–73 °C
Density:1.08 g/cm³ (20 °C)
Flash Pt:49 °C
UN:1715
ADR:8 (3),II
:102.09 g/mol
Storage Temperature:Ambient
EINECS:203-564-8
REACH:01-2119486470-36
:GHS02
:GHS07
:GHS05
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay:Min. 96 %
- Evaporation residue:Max. 100 ppm
- Heavy metals (as Pb):Max. 10 ppm
- Cl (Chloride):Max. 10 ppm
- SO4 (Sulphate):Max. 20 ppm
- Fe (Iron):Max. 10 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number Frequently Bought Together