Order Entry
Italy
ContactUsLinkComponent
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99%, Gen-Apex for molecular biology
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99%, Gen-Apex for molecular biology
Catalog # 33621.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99%, Gen-Apex for molecular biology
Catalog # 33621.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Colouration (0,5 mol/l; water):
    Max. 10 APHA
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Transmittance (230 nm) (0,5 mol/l):
    Min. 45 %
  • Transmittance (260 nm) (0,5 mol/l):
    Min. 75 %
  • Transmittance (280 nm) (0.5 mol/l):
    Min. 80 %
  • Transmittance (320 nm) (0.5 mol/l):
    Min. 90 %
  • DNases (exo- and endonucleases):
    Not detected
  • Proteases:
    Not detected
  • RNases:
    Not detected

Specification Test Results

About this item

Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
 :
 :121.14 g/mol
Merck Index:13,09842
Boiling Pt:357 °C (1013 hPa)
Melting Pt:169…172 °C
Density:1.328 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-064-4
 :GHS07
CAS Number:77-86-1
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679