Order Entry
Italy
ContactUsLinkComponent
Ethanolamine ≥99%, GPR RECTAPUR® for liquid scintillation counting
Ethanolamine ≥99%, GPR RECTAPUR® for liquid scintillation counting
Catalog # 23782.291
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ethanolamine ≥99%, GPR RECTAPUR® for liquid scintillation counting
Catalog # 23782.291
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Identification:
    Passes test
  • Boiling point:
    170 - 172 °C
  • Density (20/4):
    1.010 - 1.020
  • Solidification point:
    9.0 - 10.5 °C
  • Water:
    Max. 1 %

Specification Test Results

About this item

Synonyms: 2-Aminoethanol, Monoethanolamine
 :
 :61.08 g/mol
Melting Pt:10.5 °C
EINECS:205-483-3
UN:2491
 :GHS07
 :GHS05
Storage Temperature:Ambient
CAS Number:141-43-5
Formula:NH₂CH₂CH₂OH
MDL Number:MFCD00008183
Boiling Pt:171
Density:1.0164 g/cm³ (20 °C)
Flash Pt:93
ADR:8 III
Merck Index:13,03762