Ethanolamine, EMSURE® for analysis, Supelco®
: Merck
: 2-Aminoethanol, Monoethanolamine
:141-43-5
:
:NH₂CH₂CH₂OH
:MFCD00008183
:171
:10.5
:1.0164 g/cm³ (20 °C)
:61.08 g/mol
:93
:205-483-3
:2491
:8 III
:13,03762
:M-Clarity™ quality level = MQ300
:GHS05
:GHS07
:Ambient
- Environmentally Preferable:
- Solubility:(20 °C) soluble
- Melting Point:10.5 °C
- Molar Mass:61.08 g/mol
- Boiling Point:171 °C (1013 hPa)
- Vapor Pressure:0.5 hPa (20 °C)
- Flash Point:92.5 °C
- Refractive Index:1.4539
- Explosion Limit:3.4 - 27 %(V)
- Density:1.02 g/cm3 (20 °C)
- pH:12.1 (100 g/l, H2O, 20 °C)
- Ignition Point:410 °C
- Viscosity kinematic:20 mm2/s (23 °C)
Frequently Bought Together