Order Entry
Italy
ContactUsLinkComponent
1-(Dimethylamino)-2-propanol for synthesis, Sigma-Aldrich®
Flammable
Corrosive
Irritant
1-(Dimethylamino)-2-propanol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Corrosive
Irritant
1-(Dimethylamino)-2-propanol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -85 °C
  • Molar Mass:
    103.16 g/mol
  • Boiling Point:
    125 - 127 °C (1013 hPa)
  • Vapor Pressure:
    17.5 hPa (20 °C)
  • Flash Point:
    26 °C
  • Refractive Index:
    1.4193 (20 °C, 589 nm)
  • Explosion Limit:
    2.7 - 11.1 %(V)
  • Density:
    0.85 g/cm3 (20 °C)
  • pH:
    11 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    225 °C

Specification Test Results

Related Information

Frequently Bought Together