Thiourea for synthesis, Sigma-Aldrich®
: Merck
:62-56-6
:
:NH₂CSNH₂
:MFCD00008067
:76.12 g/mol
:13,09443
:174…177 °C
:1.435 g/cm³ (20 °C)
:Ambient
:200-543-5
:9,III
:M-Clarity™ quality level = MQ200
:3077
:GHS08
:GHS07
:GHS09
- Solubility:137 g/l (20 °C)
- Melting Point:169 - 173 °C
- Molar Mass:76.12 g/mol
- Bulk Density:640 kg/m3
- Vapor Pressure:(20 °C)
- Density:1.405 g/cm3 (20 °C)
- pH:6 - 8 (50 g/l, H2O, 20 °C)
- Ignition Point:440 °C Dust
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
Frequently Bought Together