Thioacetamide ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 62-55-5 Molecular Formula: C2H5NS Formula Weight: 75.13 Melting Point: 110-114° Solubility: Soluble in water, ethanol Beilstein Reference: 506006 Merck Reference: 14,9319 MDL No.: MFCD00008070
Synonyms: Ethanethioamide
- CAS Number:62-55-5
- Formula:CH₃CSNH₂
- MDL Number:MFCD00008070
- Molecular Weight:75.13 g/mol
- Merck Index:14,9319
- Melting Pt:110…114 °C
- Density:1.37
- Storage Temperature:Ambient
- EINECS:200-541-4
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 62-55-5 Molecular Formula: C2H5NS Formula Weight: 75.13 Melting Point: 110-114° Solubility: Soluble in water, ethanol Beilstein Reference: 506006 Merck Reference: 14,9319 MDL No.: MFCD00008070
Synonyms: Ethanethioamide
- CAS Number:62-55-5
- Formula:CH₃CSNH₂
- MDL Number:MFCD00008070
- Molecular Weight:75.13 g/mol
- Merck Index:14,9319
- Melting Pt:110…114 °C
- Density:1.37
- Storage Temperature:Ambient
- EINECS:200-541-4
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



