Order Entry
Italy
ContactUsLinkComponent
EDTA tetrasodium salt tetrahydrate 98% (by complexometric analysis), Calbiochem®
EDTA tetrasodium salt tetrahydrate 98% (by complexometric analysis), Calbiochem®
  34103-500
 :  
EDTA tetrasodium salt tetrahydrate 98% (by complexometric analysis), Calbiochem®
  34103-500
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645
 :
 :452.23 g/mol
 :12,03557
 :> 300 °C
 :6.9 g/cm³ (20 °C)
 :200-573-9
 :GHS05
 :GHS07
 :Ambient
 :13235-36-4
 :C₁₀H₁₂N₂O₈Na₄·4H₂O
 :MFCD00150027