Order Entry
Italy
ContactUsLinkComponent
Citric acid, BAKER ANALYZED® ACS, J.T.Baker®
Irritant
Citric acid, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0090.1000
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Citric acid, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0090.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    min. 99.5 %
  • Chloride (Cl):
    max. 0.001 %
  • Insoluble Matter:
    max. 0.005 %
  • Oxalate (C2O4):
    passes test
  • Phosphate (PO4):
    max. 0.001 %
  • Residue after Ignition:
    max. 0.02 %
  • Sulfur Compounds (as SO4):
    max. 0.002 %
  • Substances Carbonizable by Hot H2SO4 (Tartrates, etc.):
    passes test
  • Trace Impurities (in ppm)Iron (Fe):
    max. 3
  • Trace Impurities (in ppm)Lead (Pb):
    max. 2

Specification Test Results

About this item

 :
 :192.13 g/mol
Merck Index:13,02350
Boiling Pt:310 °C (1013 hPa)
Melting Pt:149…151 °C
Density:1.665 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-069-1
REACH:01-2119457026-42-0000
 :GHS07
CAS Number:77-92-9
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669