Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Disodium oxalate ≥99%

Disodium oxalate ≥99%
Certificates
Disodium oxalate ≥99%
Disodium oxalate ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 62-76-0 Molecular Formula: Na2C2O4 Formula Weight: 134.00 Odour: Odourless Melting Point: ca 250-270° dec. Beilstein Reference: 3631622 Merck Reference: 14,8650 MDL No.: MFCD00012465

Synonyms:  Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances
  • CAS Number:62-76-0
  • Formula:Na₂C₂O₄
  • MDL Number:MFCD00012465
  • Molecular Weight:134 g/mol
  • Merck Index:14,8650
  • Melting Pt:∼ 250…270 °C (dec.)
  • Density:2.340
  • Storage Temperature:Ambient
  • EINECS:200-550-3
Product Family Options

Product Information