
di-Potassium oxalate monohydrate 99+% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6487-48-5
:
Formula:(COOK)₂·H₂O
MDL Number:MFCD00150033
:184.23 g/mol
Merck Index:12,07820
Melting Pt:356 °C
Density:2,154 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-506-8
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White Crystals or flakes
- Infrared spectrum:Conforms
- pH:7 to 8.5 (5 % at 20°C)
- Titration with HClO4:≥99.0 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤100 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤20 ppm
Specification Test Results
Frequently Bought Together