Order Entry
Italy
ContactUsLinkComponent
Adipic acid, EMPROVE® ESSENTIAL Ph. Eur., NF, FCC, E355, SAFC®
Irritant
Adipic acid, EMPROVE® ESSENTIAL Ph. Eur., NF, FCC, E355, SAFC®
Catalog # 1.00090.9029
CAS Number:  
undefined
Irritant
Adipic acid, EMPROVE® ESSENTIAL Ph. Eur., NF, FCC, E355, SAFC®
Catalog # 1.00090.9029
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
Merck Index:13,00163
Boiling Pt:337 °C (1013 hPa)
Melting Pt:151 °C
Density:1.36 g/cm³ (25 °C)
Flash Pt:196 °C (closed cup)
 :146.14 g/mol
Storage Temperature:Ambient
EINECS:204-673-3
 :GHS07
CAS Number:124-04-9
Formula:HOOC(CH₂)₄COOH
MDL Number:MFCD00004420