

Adipic acid 99.6-101.0% (dried basis), meets analytical specification of Ph. Eur., BP, E355, Fluka™
Catalog # 27635-25KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Fiberboard box
- Pk:25 kg
- Seq:4283
Specifications
Specification Test Results
- Assay (calculated to the dried substance):99.6 - 101.0 %
- Loss on drying (105°C):Max. 0.2 %
- Water (Karl Fischer):Max. 0.2 %
- Sulfated ash:Max. 20 ppm
- Arsenic (As):Max. 3 ppm
- Iron (Fe):Max. 5 ppm
- Mercury (Hg):Max. 1 ppm
- Lead (Pb):Max. 2 ppm
- Heavy metals (as Pb):Max. 10 ppm
- Chloride (Cl):Max. 50 ppm
- Nitrate (NO₃):Max. 30 ppm
- Sulfate (SO₄):Max. 500 ppm
- Appearance of the solution:complying
- Related substances (HPLC):complying
- Residual solvents:complying
- Ignition Residue:≤0.002% (as SO₄)
- Vapor Density:5 (vs air)
- Autoignition Temperature:788 °F
Specification Test Results
About this item
:
Merck Index:13,00163
Boiling Pt:265 °C (133 hPa)
Melting Pt:151-154 °C
Melting Pt:151…154 °C
Density:1.37 g/cm³ (20 °C)
Flash Pt:196 °C
:146.14 g/mol
EINECS:204-673-3
:GHS07
:GHS05
Storage Temperature:Ambient
CAS Number:124-04-9
Formula:HOOC(CH₂)₄COOH
MDL Number:MFCD00004420