Order Entry
Italy
Orders LinkContactUsLinkComponent
Adipic acid 99%

Adipic acid 99%

Adipic acid 99%
Certificates
Adipic acid 99%

4 Options Available Below

Product Details & Documents
Synonyms:  Hexanedioc acid, 1,6-Hexanedioic acid
  • CAS Number:124-04-9
  • Formula:HOOC(CH₂)₄COOH
  • MDL Number:MFCD00004420
  • Molecular Weight:146.14 g/mol
  • Merck Index:13,00163
  • Boiling Pt:337 °C (760 mmHg)
  • Melting Pt:151…153 °C
  • Density:1,36 g/cm³ (25 °C)
  • Flash Pt:196 °C
  • Storage Temperature:Ambient
  • EINECS:204-673-3

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    151 to 153 °C
  • Water:
    ≤0.2 %
  • Ash:
    ≤0.0002 %
  • Titration with NaOH:
    ≥99 %
  • Color of solution:
    ≤5 APHA (20g adipic acid + 20 g dist. H2O + 60 g 10% NH3 soln.)
  • Nitrate (NO3):
    ≤4 ppm
  • Iron (Fe):
    ≤0.4 ppm
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.