Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Malonic acid 99%

Malonic acid 99%
 
Malonic acid 99%
Malonic acid 99%

 

 
 :  1,3-Propanedioic acid
  •  :141-82-2
  •  :C₃H₄O₄
  •  :MFCD00002707
  •  :104.06 g/mol
  •  :13,05732
  •  :387 °C (1013 hPa)
  •  :130…135 °C (dec.)
  •  :1,669 g/cm³ (20 °C)
  •  :172 °C
  •  :Ambient
  •  :205-503-0

 

 

 

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    133 to 136 °C
  • Titration with NaOH:
    ≥98.5 %
  • Chloride (Cl):
    ≤500 ppm
  • Sulfate (SO4):
    ≤500 ppm
Product Family Options

 

  • Pack type
    Pk
     
     
  • Plastic bottle
    250 g
       
       
    1 * 250 g
    1 * 250 g - 99,20 €
    99,20 €
    1 * 250 g
       99,20 €

  • Plastic bottle
    1 kg
       
       
    1 * 1 kg
    1 * 1 kg - 272,00 €
    272,00 €
    1 * 1 kg
       272,00 €

  • Plastic bottle
    5 kg
       
       
    1 * 5 kg
    1 * 5 kg - 926,00 €
    926,00 €
    1 * 5 kg
       926,00 €