Order Entry
Italy
ContactUsLinkComponent
Mercaptoacetic acid (thioglycolic acid) Reag. Ph. Eur. analytical reagent, Supelco®
Toxic
Corrosive
Mercaptoacetic acid (thioglycolic acid) Reag. Ph. Eur. analytical reagent, Supelco®
Supplier:  Merck
Certificates
undefined
Toxic
Corrosive
Mercaptoacetic acid (thioglycolic acid) Reag. Ph. Eur. analytical reagent, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -16.5 °C
  • Molar Mass:
    92.11 g/mol
  • Boiling Point:
    220 °C (1013 hPa) (decomposition)
  • Vapor Pressure:
    0.1 hPa (20 °C)
  • Flash Point:
    126 °C
  • Density:
    1.325 g/cm3 (20 °C)
  • pH:
    1.5 (10 g/l, H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive material

Specification Test Results

Related Information

Frequently Bought Together