3-Mercaptopropionic acid ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 107-96-0 Molecular Formula: C3H6O2S Formula Weight: 106.14 Flash Point: 93°(199°F) Refractive Index: 1.4920 Melting Point: 17-19° Boiling Point: 110-111°/15mm Beilstein Reference: 773807 UN No.: 2922 MDL No.: MFCD00004897
- CAS Number:107-96-0
- Formula:C₃H₆O₂S
- MDL Number:MFCD00004897
- Molecular Weight:106.15 g/mol
- Boiling Pt:110…111 °C (15 mmHg)
- Melting Pt:17…19 °C
- Density:1.220
- Flash Pt:93 °C (199 °F)
- Storage Temperature:Ambient
- EINECS:203-537-0
- UN:2922
- ADR:8,II
2 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 107-96-0 Molecular Formula: C3H6O2S Formula Weight: 106.14 Flash Point: 93°(199°F) Refractive Index: 1.4920 Melting Point: 17-19° Boiling Point: 110-111°/15mm Beilstein Reference: 773807 UN No.: 2922 MDL No.: MFCD00004897
- CAS Number:107-96-0
- Formula:C₃H₆O₂S
- MDL Number:MFCD00004897
- Molecular Weight:106.15 g/mol
- Boiling Pt:110…111 °C (15 mmHg)
- Melting Pt:17…19 °C
- Density:1.220
- Flash Pt:93 °C (199 °F)
- Storage Temperature:Ambient
- EINECS:203-537-0
- UN:2922
- ADR:8,II
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



