3-Mercaptopropionic acid 99+%
Nome del fornitore: Thermo Fisher Scientific
About this item
- CAS Number:107-96-0
- Formula:C₃H₆O₂S
- MDL Number:MFCD00004897
- Molecular Weight:106.15 g/mol
- Boiling Pt:110…111 °C (15 mmHg)
- Melting Pt:17…19 °C
- Density:1.218 g/cm³
- Flash Pt:124 °C
- Storage Temperature:Ambient
- EINECS:203-537-0
- UN:2922
- ADR:8,II
3 Options Available Below
Product Details & Documents
- CAS Number:107-96-0
- Formula:C₃H₆O₂S
- MDL Number:MFCD00004897
- Molecular Weight:106.15 g/mol
- Boiling Pt:110…111 °C (15 mmHg)
- Melting Pt:17…19 °C
- Density:1.218 g/cm³
- Flash Pt:124 °C
- Storage Temperature:Ambient
- EINECS:203-537-0
- UN:2922
- ADR:8,II
Documents
ACRO12553
Scheda di sicurezza
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4910 to 1.4940 (20°C, 589 nm)
- Titration Iodimetric:99.0 to 101.0 %



