Order Entry
Italy
ContactUsLinkComponent
L-Methionine, Millipore®
L-Methionine, Millipore®
Catalog # 4500-100
CAS Number:  
L-Methionine, Millipore®
Catalog # 4500-100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :149.21 g/mol
Merck Index:13,06004
Boiling Pt:278 °C (1013 hPa)
Melting Pt:136 °C
Density:1.373 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-562-9
CAS Number:63-68-3
Formula:C₅H₁₁NO₂S
MDL Number:MFCD00063097