Order Entry
Italy
Orders LinkContactUsLinkComponent
Lithium stearate

Lithium stearate

Glass bottle for solids, 250 g
Catalog # 39335.30
Lithium stearate
Catalog # 39335.30
CAS Number:  4485-12-5
Lithium stearate
Lithium stearate

Glass bottle for solids, 250 g

CAS Number:  4485-12-5
Price
57,70 €
Log in for availability
1 * 250 g
Product Details & Documents

CAS No.: 4485-12-5 Molecular Formula: CH3(CH2)16CO2Li Formula Weight: 290.41 Physical Form: Powder Melting Point: 220° MDL No.: MFCD00042032

Synonyms:  Stearic acid lithium salt, Lithium Stearate
  • CAS Number:4485-12-5
  • Formula:C₁₈H₃₅LiO₂
  • MDL Number:MFCD00042032
  • Molecular Weight:290.42 g/mol
  • Melting Pt:220 °C
  • Storage Temperature:Ambient
  • EINECS:224-772-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.