Order Entry
Italy
ContactUsLinkComponent
Dimethyl oxalate for synthesis, Sigma-Aldrich®
Irritant
Dimethyl oxalate for synthesis, Sigma-Aldrich®
Catalog # 8.22115.9035
Supplier:  Merck
CAS Number:  
Irritant
Dimethyl oxalate for synthesis, Sigma-Aldrich®
Catalog # 8.22115.9035
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    35 kg
  • Seq:
    4343

Specifications

Specification Test Results

  • Solubility:
    60 g/l (25 °C)
  • Melting Point:
    50 - 53 °C
  • Molar Mass:
    118.08 g/mol
  • Boiling Point:
    163 - 164 °C (1013 hPa)
  • Vapor Pressure:
    3 hPa (20 °C)
  • Flash Point:
    75 °C
  • Density:
    1.15 g/cm3 (54 °C)
  • pH:
    1 (60 g/l, H2O, 20 °C)
  • Ignition Point:
    480 °C

Specification Test Results

About this item

 :
 :118.09 g/mol
Merck Index:12,06181
Boiling Pt:163 °C (1013 hPa)
Melting Pt:54 °C
Density:1.422 g/cm³ (15 °C)
Flash Pt:62 °C
Storage Temperature:Ambient
EINECS:209-053-6
UN:3261
ADR:8,III
 :M-Clarity™ quality level = MQ200
 :GHS07
CAS Number:553-90-2
Formula:C₄H₆O₄
MDL Number:MFCD00008442