Order Entry
Italy
Orders LinkContactUsLinkComponent
Dimethyl adipate 99+%

Dimethyl adipate 99+%

Dimethyl adipate 99+%
Certificates
Dimethyl adipate 99+%

2 Options Available Below

Product Details & Documents
Synonyms:  Adipic acid dimethyl ester, 1,6-Hexanedioic acid dimethyl ester
  • CAS Number:627-93-0
  • Formula:C₈H₁₄O₄
  • MDL Number:MFCD00008469
  • Molecular Weight:174.2 g/mol
  • Merck Index:13,00163
  • Boiling Pt:109…110 °C (14 mmHg)
  • Melting Pt:8 °C
  • Density:1.06 g/cm³
  • Flash Pt:107 °C
  • Storage Temperature:Ambient
  • EINECS:211-020-6

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4270 to 1.4290 (20°C, 589 nm)
  • Water:
    ≤0.1 % (K.F.)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤20 APHA
  • Acidity:
    ≤0.01 % (as adipic acid)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 19,80 €
    19,80 €
    1 * 100 g
       19,80 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 46,60 €
    46,60 €
    1 * 500 g
       46,60 €

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..