Order Entry
Italy
ContactUsLinkComponent
 
Diethyl sebacate for synthesis, Sigma-Aldrich®
  8.00757.0250
 :  Merck
 :  
Diethyl sebacate for synthesis, Sigma-Aldrich®
  8.00757.0250
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

 

 

  • Solubility:
    0.08 g/l (20 °C)
  • Melting Point:
    -3 °C
  • Molar Mass:
    258.35 g/mol
  • Boiling Point:
    312 °C (1013 hPa)
  • Flash Point:
    71 °C
  • Density:
    0.97 g/cm3 (20 °C)
  • Viscosity kinematic:
    6.2 mm2/s (20 °C)
  • Synonyms:
    Sebacic acid diethyl ester, Decanedioic acid diethyl ester
  • Hill Formula:
    C14H26O4
  • Chemical Formula:
    C2H5OOC(CH2)8COOC2H5

 

 

 :
 :258.36 g/mol
 :312 °C (1013 hPa)
 :–3 °C
 :0.9646 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :Ambient
 :203-764-5
 :110-40-7
 :C₁₄H₂₆O₄
 :MFCD00009219