
Diethyl carbonate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:105-58-8
:
Formula:C₅H₁₀O₃
MDL Number:MFCD00009107
:118.13 g/mol
Merck Index:13,03815
Boiling Pt:127 °C (1013 hPa)
Melting Pt:–43 °C
Density:0.9751 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-311-1
UN:2366
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:33 °C
:GHS02
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:-43 °C
- Molar Mass:118.13 g/mol
- Boiling Point:126 °C (1013 hPa)
- Vapor Pressure:11 hPa (20 °C)
- Flash Point:33 °C
- Refractive Index:1.384 (20 °C)
- Explosion Limit:1.4 - 11.0 %(V)
- Density:0.97 g/cm3 (20 °C)
- pH:6.3 (1 g/l, H2O)
- Ignition Point:445 °C
- Evaporation number:14
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Specification Test Results
Frequently Bought Together