Order Entry
Italy
ContactUsLinkComponent
Methyl myristate for synthesis, Sigma-Aldrich®
Methyl myristate for synthesis, Sigma-Aldrich®
Catalog # 8.18613.0250
Supplier:  Merck
CAS Number:  
Methyl myristate for synthesis, Sigma-Aldrich®
Catalog # 8.18613.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 ml
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    18 °C
  • Molar Mass:
    242.4 g/mol
  • Boiling Point:
    162 °C (17.3 hPa)
  • Vapor Pressure:
    0.065 Pa (25 °C)
  • Flash Point:
    112 °C
  • Density:
    0.867 g/cm3 (20 °C)

Specification Test Results

About this item

Synonyms: Tetradecylic acid methyl ester, Myristic acid methyl ester, Tetradecanoic acid methyl ester, Methyl tetradecanoate
 :
 :242.4 g/mol
Boiling Pt:323
Melting Pt:18
Density:0.86715 g/cm³ (20 °C)
Flash Pt:140
Storage Temperature:Refrigerator
EINECS:204-680-1
CAS Number:124-10-7
Formula:C₁₅H₃₀O₂
MDL Number:MFCD00008983
 :M-Clarity™ quality level = MQ200