Order Entry
Italy
ContactUsLinkComponent
Paracetamol
Paracetamol
Catalog # EHERC15846000
Supplier:  EHRENSTORFER
CAS Number:  
Paracetamol
Catalog # EHERC15846000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :151.16 g/mol
Merck Index:13,00048
Boiling Pt:388 °C (1013 hPa)
Melting Pt:168 °C
Density:1.34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-157-5
CAS Number:103-90-2
Formula:C₈H₉NO₂
MDL Number:MFCD00002328