Order Entry
Italy
ContactUsLinkComponent
 
Urea ≥98%, white powder for molecular biology
 
undefined
Urea ≥98%, white powder for molecular biology

 

  • Identity Test:
    Passes
  • Purity:
    >98%
  • FTIR:
    Conforms to Standard
  • Melting Point:
    130 - 140°C
  • OD260 nm (7 M aq soln):
    <0.2
  • OD280 (7M aq soln):
    <0.2
  • pH (7 M aq soln):
    7.5 - 9.5
  • PB:
    <2 ppm

 

Frequently Bought Together