Urea 98%, pearls, extra pure
:57-13-6
:
:NH₂CONH₂
:MFCD00008022
:13,09935
:197 °C (1013 hPa)
:131…135 °C
:1,34 g/cm³ (20 °C)
:60.06 g/mol
:Ambient
:200-315-5
:01-2119463277-33
- Appearance:White Pearls
- Infrared spectrum:Conforms
- Melting point:131 to 135 °C
- Assay:≥97.5 % (ex N)
- Water:≤0.4 %
- Impurity:≤1.2 % (Biuret)
Frequently Bought Together