

N,N-Dimethylacetamide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:127-19-5
:
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686
:87.12 g/mol
Merck Index:13,03254
Boiling Pt:166.1 °C (1013 hPa)
Melting Pt:–20 °C
Density:0.937 g/cm³ (20 °C)
EINECS:204-826-4
UN:2810
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:70 °C
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-20 °C
- Molar Mass:87.12 g/mol
- Boiling Point:165 - 166 °C (1013 hPa)
- Vapor Pressure:1.76 hPa (20 °C)
- Flash Point:64 °C
- Refractive Index:1.423 (20 °C, 589 nm)
- Explosion Limit:1.7 - 11.5 %(V)
- Density:0.94 g/cm3 (20 °C)
- pH:4 (200 g/l, H2O, 20 °C)
- Ignition Point:400 °C
Specification Test Results
Related Information
Frequently Bought Together