Order Entry
Italy
Orders LinkContactUsLinkComponent
 

1,1-Dimethoxyacetone ≥97%

Flammable
Irritant
1,1-Dimethoxyacetone ≥97%
Certificates
1,1-Dimethoxyacetone ≥97%
1,1-Dimethoxyacetone ≥97%

2 Options Available Below

Product Details & Documents

Purity: 97+% CAS No.: 6342-56-9 Molecular Formula: CH3COCH(OCH3)2 Formula Weight: 118.13 Flash Point: 34°(93°F) Refractive Index: 1.3980 Melting Point: -57° Boiling Point: 133-135° Beilstein Reference: 1560557 UN No.: 1993 MDL No.: MFCD00008758

Synonyms:  Pyruvaldehyde 1,1-dimethyl acetal, 1,1-Dimethoxypropan-2-one, Pyruvic aldehyde dimethyl acetal
  • CAS Number:6342-56-9
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD00008758
  • Molecular Weight:118.13 g/mol
  • Boiling Pt:133…135 °C
  • Melting Pt:–57 °C
  • Density:0.998
  • Flash Pt:34 °C (93 °F)
  • Storage Temperature:Ambient
  • EINECS:228-735-4
  • UN:1993
  • ADR:3,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.