Order Entry
Italy
Orders LinkContactUsLinkComponent
1,1-Dimethoxyacetone 98%

1,1-Dimethoxyacetone 98%

Irritant
Flammable
1,1-Dimethoxyacetone 98%
Certificates
1,1-Dimethoxyacetone 98%

2 Options Available Below

Product Details & Documents
Synonyms:  Pyruvaldehyde 1,1-dimethyl acetal, 1,1-Dimethoxypropan-2-one, Pyruvic aldehyde dimethyl acetal
  • CAS Number:6342-56-9
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD00008758
  • Molecular Weight:118.13 g/mol
  • Boiling Pt:143 °C (1013 hPa)
  • Melting Pt:–57 °C
  • Density:1 g/cm³
  • Flash Pt:37 °C
  • EINECS:228-735-4
  • UN:1224
  • ADR:3,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3970 to 1.3990 (20°C, 589 nm)
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 43,80 €
    43,80 €
    1 * 100 g
       43,80 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 150,00 €
    150,00 €
    1 * 500 g
       150,00 €