Order Entry
Italy
ContactUsLinkComponent
Sodium pyruvate ≥99%, white powder cell culture reagent
Corrosive
Sodium pyruvate ≥99%, white powder cell culture reagent
Supplier:  MP Biomedicals
Certificates
Corrosive
Sodium pyruvate ≥99%, white powder cell culture reagent
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥99%
  • IR:
    Conforms to Standard
  • pH (1% aq soln):
    5-7

Specification Test Results

Frequently Bought Together