


Di-tert-butyl pyrocarbonate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:24424-99-5
:
Formula:C₁₀H₁₈O₅
MDL Number:MFCD00008805
:218.25 g/mol
Boiling Pt:56…57 °C (.5 mmHg)
Melting Pt:22…24 °C
Density:1.02 g/cm³
Flash Pt:37 °C
Storage Temperature:Refrigerator
EINECS:246-240-1
UN:2930
ADR:6.1,II
:GHS02
:GHS05
:GHS06
Specification Test Results
- Appearance:White Low melting crystalline solid
- Infrared spectrum:Conforms
- Titration by coupling:≥96.0 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together