
Chloral hydrate, GPR RECTAPUR®
Catalog # 22682.265
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- pH (10 %):4.0 - 5.5
- Evaporation residue:Max. 0.15 %
- Heavy metals (as Pb):Max. 100 ppm
- Ignition residue (SO4):Max. 0.1 %
- Cl (Chloride):Max. 100 ppm
Specification Test Results
About this item
:
:165.4 g/mol
Merck Index:12,02113
Boiling Pt:97.5 °C (1013 hPa)
Melting Pt:52 °C
Density:1.901 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-117-5
UN:2811
ADR:6.1,III
:GHS06
CAS Number:302-17-0
Formula:Cl₃CCH(OH)₂
MDL Number:MFCD00044479