Order Entry
Italy
ContactUsLinkComponent
Dichloroacetic acid (DCA) for synthesis, Sigma-Aldrich®
Corrosive
Irritant
Environment
Dichloroacetic acid (DCA) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Irritant
Environment
Dichloroacetic acid (DCA) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -4 °C
  • Molar Mass:
    128.94 g/mol
  • Boiling Point:
    192 °C decomposes
  • Vapor Pressure:
    0.19 hPa (20 °C)
  • Flash Point:
    1.4642 (20 °C, 589 nm)
  • Density:
    1.57 g/cm3 (20 °C)
  • pH:
    1.2 (129 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together