Dichloroacetic acid (DCA) 99+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: DCA Deblock, DCA
- CAS Number:79-43-6
- Formula:C₂H₂Cl₂O₂
- MDL Number:MFCD00004223
- Molecular Weight:128.94 g/mol
- Merck Index:13,03075
- Boiling Pt:194 °C (760 mmHg)
- Melting Pt:9…11 °C
- Density:1.56 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:201-207-0
- UN:1764
- ADR:8,II
3 Options Available Below
Product Details & Documents
Synonyms: DCA Deblock, DCA
- CAS Number:79-43-6
- Formula:C₂H₂Cl₂O₂
- MDL Number:MFCD00004223
- Molecular Weight:128.94 g/mol
- Merck Index:13,03075
- Boiling Pt:194 °C (760 mmHg)
- Melting Pt:9…11 °C
- Density:1.56 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:201-207-0
- UN:1764
- ADR:8,II
Documents
ACRO11306
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4650 to 1.4670 (20°C, 589 nm)
- Color scale:≤100 APHA
- Titration with NaOH:99.0 to 101.0 %



