11-Bromo-1-undecanol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1611-56-9
:
Formula:HO(CH₂)₁₀CH₂Br
MDL Number:MFCD00004752
:251.21 g/mol
Boiling Pt:165…170 °C (1 mmHg)
Melting Pt:46…49 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:216-554-3
Related Information
Frequently Bought Together