Order Entry
Italy
ContactUsLinkComponent
1,7-Octadiene for synthesis, Sigma-Aldrich®
Flammable
Health
1,7-Octadiene for synthesis, Sigma-Aldrich®
Catalog # 8.18797.4500
Supplier:  Merck
CAS Number:  
Flammable
Health
1,7-Octadiene for synthesis, Sigma-Aldrich®
Catalog # 8.18797.4500
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    4,5 kg
  • Seq:
    4104

Specifications

Specification Test Results

  • Solubility:
    0.08 g/l (20 °C)
  • Molar Mass:
    110.20 g/mol
  • Boiling Point:
    116 - 118 °C (1013 hPa)
  • Vapor Pressure:
    20 hPa (20 °C)
  • Flash Point:
    5 °C
  • Refractive Index:
    1.4215 (20 °C, 589 nm)
  • Explosion Limit:
    0.5 - 8.8 %(V)
  • Density:
    0.73 g/cm3 (20 °C)
  • Ignition Point:
    230 °C

Specification Test Results

About this item

 :
 :110.2 g/mol
Boiling Pt:121 °C (1013 hPa)
Melting Pt:–100 °C
Density:0.7313 g/cm³ (20 °C)
Flash Pt:5 °C
Storage Temperature:Ambient
EINECS:223-054-9
UN:2309
ADR:3,II
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS08
CAS Number:3710-30-3
Formula:C₈H₁₄
MDL Number:MFCD00008668