Order Entry
Italy
ContactUsLinkComponent
Linoleic acid for synthesis, Sigma-Aldrich®
Linoleic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Linoleic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Melting Point:
    -5 °C
  • Molar Mass:
    280.45 g/mol
  • Boiling Point:
    230 °C (21 hPa)
  • Flash Point:
    110 °C
  • Density:
    0.903 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together