D-Glucono-1,5-lactone for synthesis, Sigma-Aldrich®
Catalog # 8.43794.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Hill Formula:C₆H₁₀O₆
- Molar Mass:178.14 g/mol
- Melting Point:151 - 154 °C (decomposition)
- pH value:3.6 (10 g/l, H₂O, 20 °C)
- Bulk density:750 kg/m³
- Solubility:590 g/l (25 °C) Hydrolysis
- Storage class:10 - 13 Other liquids and solids
- WGK:WGK 1 slightly water endangering
Specification Test Results
About this item
:
:178.14 g/mol
Merck Index:13,04470
Boiling Pt:385 °C (1013 hPa)
Melting Pt:153 °C
Density:1.61 g/cm³ (–5 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:202-016-5
CAS Number:90-80-2
Formula:C₆H₁₀O₆
MDL Number:MFCD00006647