

(1S,2S)-(+)-1,2-Diaminocyclohexane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21436-03-3
:
Formula:C₆H₁₄N₂
MDL Number:MFCD00062986
:114.19 g/mol
Boiling Pt:104…110 °C (40 mmHg)
Melting Pt:43…45 °C
Flash Pt:70 °C (158 °F)
Storage Temperature:Refrigerator
UN:3259
ADR:8,III
:GHS05
:GHS07
Related Information
Frequently Bought Together