Jump to:
Product Details & Documents
Synonyms: (R)-(-)-1,2-Dihydroxy propane, (R)-(-)-1,2-Propylene glykol, (R)-Propane-1,2-diol
- CAS Number:4254-14-2
- Formula:C₃H₈O₂
- MDL Number:MFCD00066248
- Molecular Weight:76.1 g/mol
Documents
msds_814224
Scheda di sicurezza
Specifications
Specifications
- Pk:1



