

(R)-(-)-1-Methoxy-2-propanol 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4984-22-9
:49840-22-9
Formula:C₄H₁₀O₂
MDL Number:MFCD01632587
:90.12 g/mol
Melting Pt:–97 °C
Flash Pt:30 °C
Storage Temperature:Ambient
UN:3092
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4020 to 1.4040 (20°C, 589 nm)
- GC:≥98.0 %
- Specific optical rotation:-20° to -24° (24°C, 589 nm) (c=10, CHCl3)
Specification Test Results
Frequently Bought Together