Order Entry
Italy
ContactUsLinkComponent
L(+)-Potassium hydrogen tartrate 99.5-100.5%, AnalaR NORMAPUR® analytical reagent
L(+)-Potassium hydrogen tartrate 99.5-100.5%, AnalaR NORMAPUR® analytical reagent
Catalog # 27058.238
Supplier:  VWR Chemicals
CAS Number:  
undefined
L(+)-Potassium hydrogen tartrate 99.5-100.5%, AnalaR NORMAPUR® analytical reagent
Catalog # 27058.238
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    99.5 - 100.5 %
  • Insolubility in water:
    Max. 50 ppm
  • Loss on drying (105°C):
    Max. 0.5 %
  • Cl (Chloride):
    Max. 0.02 %
  • SO4 (Sulphate):
    Max. 0.05 %
  • As (Arsenic):
    Max. 1 ppm
  • Ba (Barium):
    Max. 0.1 %
  • Ca (Calcium):
    Max. 0.02 %
  • Cu (Copper):
    Max. 25 ppm
  • Fe (Iron):
    Max. 20 ppm
  • Zn (Zinc):
    Max. 25 ppm

Specification Test Results

About this item

 :
 :188.18 g/mol
Merck Index:13,07697
Melting Pt:250 °C
Density:1.943 g/cm³ (20 °C)
EINECS:212-769-1
CAS Number:868-14-4
Formula:KOOC(CHOH)₂COOH
MDL Number:MFCD00065392