Order Entry
Italy
ContactUsLinkComponent
L(+)-Potassium hydrogen tartrate 99.0-101.0% Ph. Eur.
L(+)-Potassium hydrogen tartrate 99.0-101.0% Ph. Eur.
Catalog # 27057.361
Supplier:  VWR Chemicals
CAS Number:  
undefined
L(+)-Potassium hydrogen tartrate 99.0-101.0% Ph. Eur.
Catalog # 27057.361
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Assay (calculated on dried substance):
    99.0 - 101.0 %
  • Appearance:
    Conforms (see CoA/CoS)
  • Identification B:
    Passes test
  • Identification C:
    Passes test
  • Identification D:
    Passes test
  • Spec. opt. rotation (dried substance):
    8 - 9.2 °
  • Oxalic acid:
    Max. 500 ppm
  • Cl (Chloride):
    Max. 500 ppm
  • SO4 (Sulphate):
    Max. 500 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Loss on drying (105°C):
    Max. 0.5 %
  • Residual solvents:
    Passes test

Specification Test Results

About this item

 :
 :188.18 g/mol
Merck Index:13,07697
Melting Pt:250 °C
Density:1.943 g/cm³ (20 °C)
EINECS:212-769-1
CAS Number:868-14-4
Formula:KOOC(CHOH)₂COOH
MDL Number:MFCD00065392